missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, For Nitrogen (Nitrate), Certified, 2.0% (w/v) ±0.1%, LabChem™
Specifications
Specifications
| Color | Clear, blue |
| Format | Liquid |
| Packaging | Poly Bottle |
| Quantity | 1 L |
| Storage Requirements | Room Temperature |
| Type | Copper Sulfate |