missing translation for 'onlineSavingsMsg'
Learn More

Copper slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7058654
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4295909 10 g
AA4295918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4295909 Supplier Thermo Scientific Chemicals Supplier No. 4295909
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 10 g
Odor Odorless
Weight ≈0.45g/slug
Form Slug
Percent Purity 99.9999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Copper slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only