missing translation for 'onlineSavingsMsg'
Learn More

Copper shot, 8mm (0.3in) & down, ACS, 99.9+% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol

Supplier:  Thermo Scientific Chemicals 03648236

 View more versions of this product

Catalog No. AA3648236

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper
7440-50-8
8.94g/cm3
Odorless
99.9+% (Metals basis)
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
For analysis ACS
Copper colored shot
500 g
0 hPa
Shot
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
ACS

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only