missing translation for 'onlineSavingsMsg'
Learn More

Copper rod, 6.35mm (0.25in) dia, 99.9% (metals basis)

Catalog No. p-7049355
Change view
Click to view available options
Quantity:
100 cm
5 x 100 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14126BY 100 cm
AA14126CQ 5 x 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14126BY Supplier Thermo Scientific Chemicals Supplier No. 014126BY
Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 100 cm
Odor Odorless
Weight ≈2.83g/cm
Form Rod
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper rod, 6.35mm (0.25 in.) dia.
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only