missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, -150 mesh, 99.5% (metals basis)

Catalog No. aa1016036
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1016036 500 g
AA1016022 100 g
AA10160A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1016036 Supplier Thermo Scientific Chemicals Supplier No. 01016036
Only null left

Apparent density 2.6-2.8g/cm3 | CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Density 8.94g/cm3
Quantity 500 g
Mesh Size -150 Mesh
Form Powder
Assay Percent Range 99.5%, metal basis
Chemical Name or Material Copper
Hazard Category H228-H315-H319-H335-H500
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 4.1; Packaging Group: III
EINECSNumber 231-159-6
RTECSNumber GL5325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only