missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, -100 mesh, 99%

Catalog No. AA0058436
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0058436 500 g
AA00584A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AA0058436 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 00058436

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored powder
Quantity 500 g
Mesh Size -100 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99% (Metals basis)
Chemical Name or Material Copper
missing translation for 'healthHazard1' H228-H315-H319-H335-H500
missing translation for 'healthHazard3' P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-159-6
RTECSNumber GL5325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only