missing translation for 'onlineSavingsMsg'
Learn More

Copper Phosphorus shot

$227.73 - $656.70

Chemical Identifiers

MDL Number MFCD01074956
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8835736
View Documents
Thermo Scientific Chemicals
08835736
500 g
Each for $227.73
Only null left
 
AA88357A3
View Documents
Thermo Scientific Chemicals
088357A3
2 kg
Each for $656.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD01074956

Specifications

500 g
MFCD01074956
Copper Phosphorus shot
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only