missing translation for 'onlineSavingsMsg'
Learn More

Copper Nickel wire, 0.5mm (0.02in) dia

Catalog No. AA43272G9
Change view
Click to view available options
Quantity:
25 m
100 m
500 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43272G9 25 m
AA43272H4 100 m
AA43272H9 500 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43272G9 Supplier Thermo Scientific Chemicals Supplier No. 043272G9

Copper Nickel wire is employed for sea water condenser systems and in desalination plants, as well as pipe work in chemical plants. It is being increasingly employed for brake and hydraulic suspension systems and cooling systems in cars and commercial vehicles. It is commonly used for the manufacture of coins, medals and other semi valuable applications. It is also used in cooling circuits, ammunition, sea water corrosion-resistant assemblies and condenser tubes. The good resistance to biofouling and sea water corrosion resistance of the copper nickel alloys have lead to its use in applications such as cladding for ships' hulls, legs of oil rig platforms and sea water intake screens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Copper Nickel wire
Name Note 0.5mm (0.02 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Ni 55:45 wt%
Quantity 25 m
Weight ≈1.75g/m
Form Wire
MDL Number MFCD00801098
Solubility Information Insoluble in water.

RUO – Research Use Only