missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Metal, Shot, 99.9%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C134025KG
Specifications
| Copper Metal | |
| 1 | |
| Poly Bottle | |
| 63.54 | |
| Ungraded |
| 7440-50-8 | |
| 2.5 kg | |
| Shot | |
| 99.9% |
Chemical Identifiers
| 7440-50-8 | |
| 63.54 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.