missing translation for 'onlineSavingsMsg'
Learn More

Copper Metal, Shot, 99.9%, Spectrum™ Chemical

Catalog No. 18609934
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609934 125 g Poly Bottle
18609936 500 g Poly Bottle
18609935 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609934 Supplier Spectrum Chemical Mfg Cor Supplier No. C1340125GM
Only null left

Chemical Identifiers

CAS 7440-50-8
Molecular Weight (g/mol) 63.54

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Form Shot
Percent Purity 99.9%
Grade Ungraded
Chemical Name or Material Copper Metal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.