missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Metal, Light, Turnings, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Packaging | Poly Bottle |
| Form | Turnings |
| Grade | Ungraded |
| Chemical Name or Material | Copper Metal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.