missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tungsten oxide, 99.5% (metals basis)

Catalog No. AA4021730
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4021730 250 g
AA4021718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4021730 Supplier Thermo Scientific Chemicals Supplier No. 04021730

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13587-35-4

Copper(II) tungsten oxide nanopowder is used for supercapacitor applications. Nano-sized CuWO4 thin films have been fabricated by radio-frequency (R.F.) sputtering deposition, and are used as positive electrode with both LiClO4 liquid electrolyte and LiPON solid electrolyte in rechargeable lithium batteries. Thin films of copper tungstate is used for nitric oxide sensing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13587-35-4
MDL Number MFCD00016067
Synonym Copper tungstate

Specifications

Physical Form Powder
Quantity 250 g
Odor Odorless
Merck Index 14,2657
Solubility Information Insoluble in water.
Formula Weight 311.38
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Copper(II) tungsten oxide
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 237-022-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only