missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T12925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12925G 5 g
T129225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12925G Supplier TCI America Supplier No. T12925G

Chemical Identifiers

CAS 34946-82-2
Molecular Formula CHCuF3O3S
Molecular Weight (g/mol) 213.617
MDL Number MFCD00077492
InChI Key GZWXEFRPSWBAGC-UHFFFAOYSA-N
Synonym copper; trifluoromethanesulfonic acid
PubChem CID 45051791
IUPAC Name copper;trifluoromethanesulfonic acid
SMILES C(F)(F)(F)S(=O)(=O)O.[Cu]

Specifications

Quantity 5 g
UN Number 1759
Formula Weight 361.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) Trifluoromethanesulfonate