missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Trifluoromethanesulfonate, 98%

Catalog No. AC405420250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405420250 25 g Glass bottle
AC405420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405420250 Supplier Thermo Scientific Chemicals Supplier No. 405420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34946-82-2 | C2CuF6O6S2 | 361.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34946-82-2
Molecular Formula C2CuF6O6S2
Molecular Weight (g/mol) 361.69
MDL Number MFCD00077492
InChI Key GZWXEFRPSWBAGC-UHFFFAOYSA-N
Synonym copper; trifluoromethanesulfonic acid
PubChem CID 45051791
IUPAC Name copper;trifluoromethanesulfonic acid
SMILES C(F)(F)(F)S(=O)(=O)O.[Cu]

Specifications

Color Blue to White or Gray
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CF3SO3)2Cu
Quantity 25 g
Fieser 07,76; 08,126; 10,110; 14,102
Formula Weight 361.69
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) trifluoromethanesulfonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 252-300-8

RUO – Research Use Only