missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) trifluoromethanesulfonate, 98%

Catalog No. AAB2025322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2025322 100 g
AAB2025306 5 g
AAB2025314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2025322 Supplier Thermo Scientific Chemicals Supplier No. B2025322

CAS: 34946-82-2 | CHCuF3O3S | 213.617 g/mol

Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 34946-82-2
Molecular Formula CHCuF3O3S
Molecular Weight (g/mol) 213.617
MDL Number MFCD00077492
InChI Key GZWXEFRPSWBAGC-UHFFFAOYSA-N
Synonym copper; trifluoromethanesulfonic acid
PubChem CID 45051791
IUPAC Name copper;trifluoromethanesulfonic acid
SMILES C(F)(F)(F)S(=O)(=O)O.[Cu]

Specifications

Melting Point (decomposition)
Quantity 100 g
UN Number UN3261
Beilstein 4028198
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 361.68
Percent Purity 98%
Chemical Name or Material Copper(II) trifluoromethanesulfonate
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-300-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only