missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfide, ultra dry, 99.98% (metals basis)

Catalog No. p-7049415
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1430009 10 g
AA1430018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AA1430009 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 01430009
Only null left

CAS: 1317-40-4 | CuS | 95.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 1317-40-4
Molecular Formula CuS
Molecular Weight (g/mol) 95.61
MDL Number MFCD00016066
InChI Key OMZSGWSJDCOLKM-UHFFFAOYSA-N
Synonym copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
PubChem CID 14831
SMILES [S--].[Cu++]

Specifications

Melting Point 220°C (decomposition)
Physical Form Powder
Quantity 10 g
Merck Index 14,2965
IUPAC Name sulfanylidenecopper
Formula Weight 95.6
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Refractive Index 1.45
Sensitivity Air and moisture sensitive
Density 3.9 g/mL to 4.6 g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) sulfide, Ultra dry
Show More Show Less
EINECSNumber 215-271-2
RTECSNumber GL8912000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only