missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate, Pentahydrate, ACS, 99.0 to 100.5%, MilliporeSigma™

Catalog No. M1027900250
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1027900250 250 g
M1027901000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1027900250 Supplier Technidata Supplier No. 1.02790.0250

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.0 to 100.5%
Linear Formula CuSO4 · 5H2O
Formula Weight 249.68
Grade ACS/EP/ISO/Reagent
Density 2.284g/cm3 (20°C)
Chemical Name or Material Copper(II) Sulfate, Pentahydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions