missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%

Catalog No. AA43760A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43760A1 1 kg
AA43760A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43760A1 Supplier Thermo Scientific Chemicals Supplier No. 043760A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16570-28-8 | Cu2O7P2 | 301.03 g/mol

Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16570-28-8
Molecular Formula Cu2O7P2
Molecular Weight (g/mol) 301.03
MDL Number MFCD00150225
InChI Key PEVJCYPAFCUXEZ-UHFFFAOYSA-J
IUPAC Name dicopper(2+) (phosphonatooxy)phosphonate
SMILES [Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Color Blue
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 301.02 (Anhydrous)
Concentration or Composition (by Analyte or Components) P2O7 43.5 to 48.5%
Chemical Name or Material Copper (II) pyrophosphate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-250-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only