Learn More
Copper(II) pyrithione
CAS: 14915-37-8 | C10H8CuN2O2S2 | 315.86 g/mol
Supplier: Thermo Scientific Chemicals 457210050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Copper(II) pyrithione | |
| 5 g | |
| Solubility in water: insoluble. | |
| 315.86 | |
| Authentic |
| 14915-37-8 | |
| C10H8CuN2O2S2 | |
| 315.86 | |
| ≥97.5% (HPLC) |
Chemical Identifiers
| 14915-37-8 | |
| C10H8CuN2O2S2 | |
| 315.86 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated cl
GHS Signal Word: Danger
EINECSNumber : 238-984-
RUO – Research Use Only