missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) perchlorate hexahydrate, Reagent Grade

Catalog No. p-7048846
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1165118 50 g
AA1165130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1165118 Supplier Thermo Scientific Chemicals Supplier No. 01165118
Only null left

CAS: 10294-46-9 | Cl2CuH12O14 | 370.528 g/mol

Copper(II) perchlorate hexahydrate is a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-46-9
Molecular Formula Cl2CuH12O14
Molecular Weight (g/mol) 370.528
MDL Number MFCD00149666
InChI Key NHELIHXBJRANPL-UHFFFAOYSA-L
Synonym copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
PubChem CID 12846356
IUPAC Name copper;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]

Specifications

Melting Point 82°C
Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Merck Index 14,2647
Solubility Information Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
Formula Weight 370.53 (262.44 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.505
Sensitivity Hygroscopic
Density 2.23 g/mL
Chemical Name or Material Copper(II) perchlorate hexahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-391-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only