missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) nitrate trihydrate, Honeywell Fluka™
Puriss. p.a., 99-104%
$931.15 - $931.15
Chemical Identifiers
| CAS | 10031-43-3 |
|---|---|
| Molecular Formula | CuH6N2O9 |
| Molecular Weight (g/mol) | 241.599 |
| MDL Number | MFCD00149671 |
| InChI Key | SXTLQDJHRPXDSB-UHFFFAOYSA-N |
| Synonym | copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem |
| PubChem CID | 9837674 |
| IUPAC Name | copper;dinitrate;trihydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 10031-43-3 | |
| 241.599 | |
| SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
| 9837674 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
| CuH6N2O9 | |
| MFCD00149671 | |
| copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
| copper;dinitrate;trihydrate |
Specifications
| 10031-43-3 | |
| CuH6N2O9 | |
| MFCD00149671 | |
| copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] | |
| 241.599 | |
| 241.6g/mol | |
| Copper(II) nitrate trihydrate |
| 500 g | |
| Cu(NO3)2 · 3H2O | |
| UN1477 | |
| SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
| copper;dinitrate;trihydrate | |
| 9837674 | |
| 99 to 104% |
Safety and Handling
P220-P273-P280-P305 + P351 + P338
RTECSNumber : GL7875000