Learn More
Copper(II) nitrate trihydrate, 99%, pure
CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol
$158.01 - $158.01
Chemical Identifiers
| CAS | 10031-43-3 |
|---|---|
| Molecular Formula | CuN2O6·3H2O |
| Molecular Weight (g/mol) | 241.6 |
| InChI Key | SXTLQDJHRPXDSB-UHFFFAOYSA-N |
| Synonym | copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem |
| PubChem CID | 9837674 |
| IUPAC Name | copper;dinitrate;trihydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453960010
|
Thermo Scientific Chemicals
453960010 |
1 kg |
Each for $158.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10031-43-3 | |
| 241.6 | |
| copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
| copper;dinitrate;trihydrate |
| CuN2O6·3H2O | |
| SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
| 9837674 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] |
Specifications
| 114.0°C | |
| 1 kg | |
| CuN2O6·3H2O | |
| 15,2633 | |
| Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2] | |
| 241.6 | |
| 241.6 | |
| Pure |
| 10031-43-3 | |
| 98.5 to 104% (Iodometry) | |
| Cu(NO3)2·3H2O | |
| copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem | |
| SXTLQDJHRPXDSB-UHFFFAOYSA-N | |
| copper;dinitrate;trihydrate | |
| 9837674 | |
| 99% | |
| Copper(II) nitrate trihydrate |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
RUO – Research Use Only