missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)

CAS: 13778-31-9 | CuN2O6 | 187.55 g/mol

Supplier:  Thermo Scientific Chemicals 01069922

 View more versions of this product

Catalog No. AA1069922


Only null left
Explore available promotions

Description

Description

It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) nitrate hydrate
Blue
100 g
CuN2O6
copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci
XTVVROIMIGLXTD-UHFFFAOYSA-N
copper;dinitrate;hydrate
15303652
Odorless
13778-31-9
Crystalline Aggregates
99.999% (Metals basis)
MFCD00149669
Soluble in ethanol and water. Insoluble in ethyl acetate.
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
187.55
187.55 (Anhydrous)
Hygroscopic

Chemical Identifiers

13778-31-9
187.55
XTVVROIMIGLXTD-UHFFFAOYSA-N
15303652
CuN2O6
MFCD00149669
copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 221-838-5

RTECSNumber : GL7875000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only