Learn More
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Description
It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Blue |
| Physical Form | Crystalline Aggregates |
| Quantity | 500 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 99.999% (Metals basis) |
| Solubility Information | Soluble in ethanol and water. Insoluble in ethyl acetate. |
| IUPAC Name | copper;dinitrate;hydrate |
| Formula Weight | 187.55 (Anhydrous) |
| Chemical Name or Material | Copper(II) nitrate hydrate |
Safety and Handling
| Hazard Category | H272-H314-H335 |
| Precautionary Statement | P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: III |
| EINECSNumber | 221-838-5 |
| RTECSNumber | GL7875000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only