missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Nitrate Hemi(Pentahydrate), Honeywell Fluka™
≥98%,≥26% Cu basis
$131.93 - $131.93
Chemical Identifiers
| CAS | 19004-19-4 |
|---|---|
| MDL Number | MFCD02253001 |
| Synonym | dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 19004-19-4 | |
| MFCD02253001 | |
| dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate |
Specifications
| 19004-19-4 | |
| Cu(NO3)2 · 2.5H2O | |
| 3085 | |
| 232.59g/mol | |
| Plastic Bottle |
| 500 g | |
| MFCD02253001 | |
| dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate | |
| ≥98% | |
| Copper(Ii) Nitrate Hemi(Pentahydrate) |
Safety and Handling
P210-P260-P280-P305 + P351 + P338-P370 + P378-P371 + P380 + P375
H271-H290-H302-H314-H410
EINECSNumber : 221-838-5
RTECSNumber : QU7400000