missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hemi(pentahydrate), 98%

CAS: 19004-19-4 | CuN2O6·2.5H2O

$92.85 - $937.38

Chemical Identifiers

CAS 19004-19-4
Molecular Formula CuN2O6·2.5H2O
MDL Number MFCD00149671
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1400430
View Documents
Thermo Scientific Chemicals
A1400430
250 g
Each for $92.85
Only null left
 
AAA140040B
View Documents
Thermo Scientific Chemicals
A140040B
1000 g
Each for $225.96
Only null left
 
AAA140040I
View Documents
Thermo Scientific Chemicals
A140040I
5000 g
Each for $937.38
Only null left
 
Description

Description

Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

Solubility
Soluble in water, ammonia and ethanol

Notes
Incompatible with reducing agents, organic materials and powdered metals.
Specifications

Chemical Identifiers

19004-19-4
CuN2O6·2.5H2O
MFCD00149671
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

170°C (decomposition)
19004-19-4
CuN2O6·2.5H2O
MFCD00149671
14,2643
Soluble in water,ammonia and ethanol
98%
Hygroscopic
Copper(II) nitrate hemi(pentahydrate)
∼115°C
250 g
Cu(NO3)2·2.5H2O
UN3085
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate
232.59 (187.55 Anhydrous)
Odorless
2.32
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 221-838-5

RTECSNumber : GL7875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only