missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) nitrate hemi(pentahydrate), 98%
Description
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
Specifications
Specifications
| Boiling Point | 170°C (decomposition) |
| Melting Point | ∼115°C |
| Quantity | 1000 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.32 |
| Linear Formula | Cu(NO3)2·2.5H2O |
| UN Number | UN3085 |
| Merck Index | 14,2643 |
| Solubility Information | Soluble in water,ammonia and ethanol |
| Show More |
Safety and Handling
| Hazard Category | H272-H314-H335 |
| Hazard Statement | GHS H Statement H272-H314-H318-H302 May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 221-838-5 |
| RTECSNumber | GL7875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only