missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) methacrylate, tech.

Catalog No. p-7051596
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4162106 5 g
AA4162114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4162106 Supplier Thermo Scientific Chemicals Supplier No. 04162106
Only null left

CAS: 19662-59-0 | C4H5CuO2 | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19662-59-0
Molecular Formula C4H5CuO2
Molecular Weight (g/mol) 148.63
MDL Number MFCD00080547,MFCD00156451,MFCD00080547
InChI Key BFHDHRZKYIRNRP-UHFFFAOYSA-M
PubChem CID 121233728
IUPAC Name copper;2-methylprop-2-enoic acid;hydrate
SMILES [Cu++].CC(=C)C([O-])=O

Specifications

Color Blue-Green
Linear Formula [H2C=C(CH3)CO2]2Cu
Quantity 5 g
Formula Weight 233.71
Grade Technical
Physical Form Powder
Chemical Name or Material Copper(II) methacrylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-211-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only