missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)

Catalog No. AA4187004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4187004 2 g
AA4187009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4187004 Supplier Thermo Scientific Chemicals Supplier No. 04187004

CAS: 155640-85-0 | C10H2CuF12O4 | 477.65 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155640-85-0
Molecular Formula C10H2CuF12O4
Molecular Weight (g/mol) 477.65
MDL Number MFCD00151019
InChI Key HZXGNBMOOYOYIS-PAMPIZDHSA-L
Synonym copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate
PubChem CID 56845342
SMILES [Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 220°C (decomposition)
Melting Point ∼100°C
Physical Form Crystalline
Quantity 2 g
Solubility Information Soluble in methanol,acetone,toluene
IUPAC Name copper(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Formula Weight 477.64 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Copper(II) hexafluoro-2,4-pentanedionate hydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-850-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only