Learn More
Copper(II) fluoride dihydrate
Flux in casting gray iron | CAS: 13454-88-1 | CuF2H4O2 | 137.57 g/mol
$99.58 - $181.82
Chemical Identifiers
| CAS | 13454-88-1 |
|---|---|
| Molecular Formula | CuF2H4O2 |
| Molecular Weight (g/mol) | 137.57 |
| MDL Number | MFCD00010977 |
| InChI Key | CTRICEWDSISGPV-UHFFFAOYSA-L |
| Synonym | cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride |
| PubChem CID | 24884278 |
| IUPAC Name | copper;difluoride;dihydrate |
| SMILES | O.O.[F-].[F-].[Cu++] |
Description
Copper(II) fluoride can also be used to form fluorobenzene from benzene, hydrochloric acid and oxygen. Using a metal fluoride that can successfully oxidize with a methyl bond.Â
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13454-88-1 | |
| 137.57 | |
| CTRICEWDSISGPV-UHFFFAOYSA-L | |
| 24884278 | |
| O.O.[F-].[F-].[Cu++] |
| CuF2H4O2 | |
| MFCD00010977 | |
| cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride | |
| copper;difluoride;dihydrate |
Specifications
| >130°C (decomposition) | |
| Powder | |
| CuF2H4O2 | |
| UN3288 | |
| cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride | |
| CTRICEWDSISGPV-UHFFFAOYSA-L | |
| copper;difluoride;dihydrate | |
| 24884278 | |
| Odorless | |
| Copper(II) fluoride dihydrate |
| 13454-88-1 | |
| 50 g | |
| MFCD00010977 | |
| 14,2638 | |
| Slightly soluble in cold water, hydrolyzes to oxyfluoride in hot water | |
| O.O.[F-].[F-].[Cu++] | |
| 137.57 | |
| 137.57 (101.54 Anhydrous) | |
| 2.934 g/mL |
Safety and Handling
GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 232-147-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only