missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride, anhydrous, 99.5%

Catalog No. p-7048792
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1148909 10 g
AA1148918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1148909 Supplier Thermo Scientific Chemicals Supplier No. 01148909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent | CAS: 7789-19-7

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent.Copper(II) fluoride is used in the preparation of fluorobenzene by reacting with benzene. It is also used in galvanic cells, high-temperature fluorinating agent and fluxing iron castings. Further, it is used as an efficient catalyst in Chan-Lam-Evans coupling reaction to synthesis the diaryl- ether compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-19-7
MDL Number MFCD00010977

Specifications

Melting Point 950°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,2638
Solubility Information Solubility in water at 20° ≈ 4;7g/100mL
Formula Weight 101.54
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 4.85 g/mL
Chemical Name or Material Copper(II) fluoride, Anhydrous
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-147-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only