missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) cyclohexanebutyrate, AAS, Cu 15.8%

Catalog No. p-7080361
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8726203 1 g
AA8726206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8726203 Supplier Thermo Scientific Chemicals Supplier No. 08726203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2218-80-6 | C20H36CuO4 | 404.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2218-80-6
Molecular Formula C20H36CuO4
Molecular Weight (g/mol) 404.05
MDL Number MFCD00036399
InChI Key ZGMZTUJLZVTXNR-UHFFFAOYSA-N
Synonym cyclohexanebutyric acid, copper salt
PubChem CID 131876184
IUPAC Name copper;4-cyclohexylbutanoic acid
SMILES C1CCC(CC1)CCCC(=O)O.C1CCC(CC1)CCCC(=O)O.[Cu]

Specifications

Melting Point 126°C (decomposition)
Quantity 1 g
Formula Weight 402.01
Grade AAS
Physical Form Powder
Chemical Name or Material Copper (II) cyclohexanebutyrate
EINECSNumber 218-723-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only