Learn More
Copper(II) chloride dihydrate, 99%, extra pure, powder
CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol
Supplier: Thermo Scientific Chemicals 405840025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Copper(II) chloride dihydrate | |
| 99% | |
| White | |
| 2.5 kg | |
| Cl2CuH4O2 | |
| MFCD00149674 | |
| 15, 2623 | |
| Solubility in water: 1150g/L (20°C). Other solubilities: soluble in alcohol,moderately soluble in acetone and ethylacetate,slightly soluble in ether | |
| O.O.[Cl-].[Cl-].[Cu++] | |
| 170.48 | |
| CHEBI:86318 | |
| 99% | |
| Glass bottle |
| 598°C | |
| 10125-13-0 | |
| Crystalline Powder | |
| 98.5% min. (Iodometry) | |
| CuCl2·2H2O | |
| 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152 | |
| coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
| MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
| dichlorocopper;dihydrate | |
| 61482 | |
| 170.48 | |
| Extra Pure | |
| 2.54 |
Chemical Identifiers
| 10125-13-0 | |
| 170.48 | |
| MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
| 61482 | |
| O.O.[Cl-].[Cl-].[Cu++] |
| Cl2CuH4O2 | |
| MFCD00149674 | |
| coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701 | |
| CHEBI:86318 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
RTECSNumber : GL7030000
RUO – Research Use Only