missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) chloride dihydrate, 99%, extra pure, powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 598°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Packaging | Glass bottle |
| Assay Percent Range | 98.5% min. (Iodometry) |
| Density | 2.54 |
| Linear Formula | CuCl2·2H2O |
| Fieser | 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152 |
| Merck Index | 15, 2623 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes serious eye damage. Causes skin irritation. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minut |
| RTECSNumber | GL7030000 |
RUO – Research Use Only