missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride dihydrate, 99+%, ACS reagent

Catalog No. AC315281000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315281000 100 g Plastic bottle
AC315285000 500 g Plastic bottle
AC315280025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315281000 Supplier Thermo Scientific Chemicals Supplier No. 315281000

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name dichlorocopper;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 100.0°C
Color Colorless
Physical Form Liquid
Quantity 100 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula CuCl2·2H2O
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152
Merck Index 15,2623
Solubility Information Solubility in water: 1150g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in acetone and ethylacetate, slightly soluble in ether
Formula Weight 170.48
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Copper(II) chloride dihydrate, For analysis ACS
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

RUO – Research Use Only