missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Chloride Dihydrate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Catalog No. p-7120348
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045851 100 g Poly Bottle
60045852 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045851 Supplier Honeywell Chemicals Supplier No. 307483100G
Only null left

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Boiling Point 993°C
Melting Point 598°C
Quantity 100 g
Linear Formula CuCl2 · 2H2O
UN Number UN2802
IUPAC Name dichlorocopper;dihydrate
Formula Weight 170.48g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.53g/cm3 Vapor Density: >1 (vs air)
Chemical Name or Material Copper(Ii) Chloride Dihydrate
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338
Precautionary Statement H290-H302 + H312-H315-H318-H410
DOTInformation 8/PGIII
EINECSNumber 231-210-2
RTECSNumber GL7030000