missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride, anhydrous, 98% min

Catalog No. p-4407437
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1245718 50 g
AA1245730 250 g
AA12457A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1245718 Supplier Thermo Scientific Chemicals Supplier No. 01245718
Only null left

For Isomerization and Cracking Catalyst, Mordant in Dyeing and Printing Fabrics, in Electroplating Baths For Plating Cu On Al | CAS: 7447-39-4 | CuCl2

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7447-39-4
Molecular Formula CuCl2
MDL Number MFCD00010972

Specifications

Boiling Point 993°C (decomposition)
Melting Point 620°C
Color Brown-Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,alcohol,acetone and hot sulfuric acid.
Formula Weight 134.45
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow-brown powder
Density 3.386 g/cm3
Chemical Name or Material Copper(II) chloride, Anhydrous
Show More Show Less
Hazard Category H302+H312-H315-H318
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-210-2
RTECSNumber GL7000000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only