missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) carbonate dihydroxide, Cu 55% min

Catalog No. AA3330536
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3330536 500 g
AA33305A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3330536 Supplier Thermo Scientific Chemicals Supplier No. 03330536

CAS: 12069-69-1 | CH2Cu2O5

Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CH2Cu2O5
MDL Number MFCD00010976
Synonym Copper(II) carbonate, basic, Copper(II) hydroxide carbonate

Specifications

Melting Point 200°C (decomposition)
Color Green
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in acid,ammonia,ammonium salt and alkali metal carbonate solution. Insoluble in water and alcohol.
Formula Weight 221.11
Density 4
Chemical Name or Material Copper(II) carbonate dihydroxide, Cu 55% min.
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 235-113-6
RTECSNumber GL6910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only