missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) carbonate basic, Solstice
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 250 g |
| Linear Formula | CuCO3 · Cu(OH)2 |
| UN Number | NONH for all modes of transport |
| Molecular Weight (g/mol) | 221.12 |
| Formula Weight | 221.12g/mol |
| Percent Purity | ≥95% (RT) |
| Chemical Name or Material | Copper(II) carbonate basic |
Safety and Handling
| Hazard Statement | P261-P305 + P351 + P338 |
| RTECSNumber | GL6910000 |