Learn More
Copper(II) acetate, anhydrous, 98%
CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol
Supplier: Thermo Scientific Chemicals B2361522
Description
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCopper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.
Solubility
Slightly soluble in water. Soluble in alcohol and ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
| Copper(II) acetate | |
| 142-71-2 | |
| Powder | |
| 98% | |
| MFCD00008690 | |
| Slightly soluble in water. Soluble in alcohol and ether. | |
| CC(=O)[O-].CC(=O)[O-].[Cu+2] | |
| 181.634 | |
| 181.63 | |
| Hygroscopic |
| Anhydrous | |
| Blue-Green | |
| 100 g | |
| C4H6CuO4 | |
| copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus | |
| OPQARKPSCNTWTJ-UHFFFAOYSA-L | |
| copper;diacetate | |
| 8895 | |
| Odorless |
Chemical Identifiers
| 142-71-2 | |
| 181.634 | |
| OPQARKPSCNTWTJ-UHFFFAOYSA-L | |
| 8895 | |
| CC(=O)[O-].CC(=O)[O-].[Cu+2] |
| C4H6CuO4 | |
| MFCD00008690 | |
| copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus | |
| copper;diacetate |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335-H373
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 205-553-3
RTECSNumber : AG3480000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only