missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate, 99.999% (metals basis)

CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol

Supplier:  Thermo Scientific Chemicals 04435514

 View more versions of this product

Catalog No. AA4435514


Only null left
Explore available promotions

Description

Description

Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) acetate
Green
25 g
C4H6CuO4
copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus
OPQARKPSCNTWTJ-UHFFFAOYSA-L
copper;diacetate
8895
Odorless
1.93g/cm3
142-71-2
Powder
99.999% (Metals basis)
MFCD00008690
Soluble in water and alcohol. Slightly soluble in ether and glycerol.
CC(=O)[O-].CC(=O)[O-].[Cu+2]
181.634
181.63
Hygroscopic

Chemical Identifiers

142-71-2
181.634
OPQARKPSCNTWTJ-UHFFFAOYSA-L
8895
CC(=O)[O-].CC(=O)[O-].[Cu+2]
C4H6CuO4
MFCD00008690
copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus
copper;diacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335-H373

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 205-553-3

RTECSNumber : AG3480000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only