missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate, 99.999% (metals basis)

Catalog No. AA4435506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4435506 5 g
AA4435514 25 g
AA4435522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4435506 Supplier Thermo Scientific Chemicals Supplier No. 04435506
Only null left

CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol

Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142-71-2
Molecular Formula C4H6CuO4
Molecular Weight (g/mol) 181.634
MDL Number MFCD00008690
InChI Key OPQARKPSCNTWTJ-UHFFFAOYSA-L
Synonym copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus
PubChem CID 8895
IUPAC Name copper;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Cu+2]

Specifications

Color Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water and alcohol. Slightly soluble in ether and glycerol.
Formula Weight 181.63
Odor Odorless
Sensitivity Hygroscopic
Density 1.93g/cm3
Chemical Name or Material Copper(II) acetate
Hazard Category H302-H314-H335-H373
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 205-553-3
RTECSNumber AG3480000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only