missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine 97.0+%, TCI America™

Catalog No. C3051200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3051200MG 200 mg
C30511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3051200MG Supplier TCI America Supplier No. C3051200MG

Chemical Identifiers

CAS 155773-67-4
Molecular Formula C80H88CuN8O8
Molecular Weight (g/mol) 1353.178
MDL Number MFCD00191950
InChI Key JJIWQZDKAOMTKU-UHFFFAOYSA-N
Synonym 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Copper(II)
PubChem CID 22834032
SMILES CCCCOC1=C2C=CC=CC2=C(C3=C4[N-]C(=C31)N=C5C6=C(C7=CC=CC=C7C(=C6C(=NC8=NC(=NC9=NC(=N4)C1=C(C2=CC=CC=C2C(=C19)OCCCC)OCCCC)C1=C(C2=CC=CC=C2C(=C18)OCCCC)OCCCC)[N-]5)OCCCC)OCCCC)OCCCC.[Cu+2]

Specifications

Quantity 200 mg
Formula Weight 1353.18
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine