missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2-ethylhexanoate

Catalog No. p-7049734
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2053909 10 g
AA2053918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2053909 Supplier Thermo Scientific Chemicals Supplier No. 02053909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149-11-1 | C16H32CuO4 | 351.974 g/mol

Copper(II) 2-ethylhexanoate is used as pharmaceutical intermediates. Used as drier/additive for paints, coatings and inks. Used as catalystic special applications in lubricant and grease additives, fuel combustion additives to enhance and stabilize the performance of fuels. Used in the the rubber wire coat compound and the brass-coated steel belt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149-11-1
Molecular Formula C16H32CuO4
Molecular Weight (g/mol) 351.974
MDL Number MFCD00015695
InChI Key SZXCZUDYSKBGSX-UHFFFAOYSA-N
Synonym copper bis 2-ethylhexanoate
PubChem CID 85309523
IUPAC Name copper;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Cu]

Specifications

Melting Point 252°C (decomposition)
Color Blue-Green
Quantity 10 g
Solubility Information Soluble in water,alcohols,dichloroethane,chlorobenzene,and most organic solvents.
Formula Weight 349.96
Physical Form Powder or Lumps
Chemical Name or Material Copper(II) 2-ethylhexanoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-731-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only