missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Trifluoromethanethiolate 90.0+%, TCI America™

$324.35 - $1012.17

Chemical Identifiers

CAS 3872-23-9
Molecular Formula CHCuF3S
Molecular Weight (g/mol) 165.62
MDL Number MFCD00059046
InChI Key JHTKDYFNOCMNDU-UHFFFAOYSA-N
Synonym (Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt
PubChem CID 53444602
IUPAC Name copper;trifluoromethanethiol
SMILES C(F)(F)(F)S.[Cu]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C11591G
View Documents
TCI America
C11591G
1 g
Each for $324.35
Only null left
 
C11595G
View Documents
TCI America
C11595G
5 g
Each for $1,012.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3872-23-9
165.62
JHTKDYFNOCMNDU-UHFFFAOYSA-N
53444602
C(F)(F)(F)S.[Cu]
CHCuF3S
MFCD00059046
(Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt
copper;trifluoromethanethiol

Specifications

3872-23-9
MFCD00059046
(Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt
C(F)(F)(F)S.[Cu]
165.62
164.61
Crystalline Powder
CHCuF3S
1 g
JHTKDYFNOCMNDU-UHFFFAOYSA-N
copper;trifluoromethanethiol
53444602
≥90.0% (T)
Copper(I) Trifluoromethanethiolate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'