missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Trifluoromethanethiolate 90.0+%, TCI America™

Catalog No. C11595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11595G 5 g
C11591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11595G Supplier TCI America Supplier No. C11595G
Only null left

Chemical Identifiers

CAS 3872-23-9
Molecular Formula CHCuF3S
Molecular Weight (g/mol) 165.62
MDL Number MFCD00059046
InChI Key JHTKDYFNOCMNDU-UHFFFAOYSA-N
Synonym (Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt
PubChem CID 53444602
IUPAC Name copper;trifluoromethanethiol
SMILES C(F)(F)(F)S.[Cu]

Specifications

Quantity 5 g
Formula Weight 164.61
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(I) Trifluoromethanethiolate
TSCA No
Recommended Storage Refrigerator