missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Trifluoromethanethiolate 90.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 164.61 |
| Percent Purity | ≥90.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Copper(I) Trifluoromethanethiolate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |