missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Trifluoromethanesulfonate Benzene Complex 90.0+%, TCI America™

Catalog No. T14421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14421G 1 g
T14425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14421G Supplier TCI America Supplier No. T14421G

Chemical Identifiers

CAS 42152-46-5
Molecular Formula CCuF3O3S
MDL Number MFCD00043300
Synonym Copper(I) Triflate Benzene Complex, Trifluoromethanesulfonic Acid Copper(I) Salt Benzene Complex

Specifications

Melting Point 160°C
Quantity 1 g
UN Number 3175
Formula Weight 251.67
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(I) Trifluoromethanesulfonate Benzene Complex
TSCA No
Recommended Storage Refrigerator