missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) trifluoromethanesulfonate benzene complex, 90%

Catalog No. p-2691239
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC371090010 1 g Glass bottle
AC371090050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC371090010 Supplier Thermo Scientific Chemicals Supplier No. 371090010

CAS: 42152-46-5 | C8H6Cu2F6O6S2 | 503.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42152-46-5
Molecular Formula C8H6Cu2F6O6S2
Molecular Weight (g/mol) 503.33
MDL Number MFCD00043300
Synonym Trifluoromethanesulfonic acid, copper(I) salt, benzene complex

Specifications

Melting Point 160.0°C
Color Gray-Green
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula (CF3SO3Cu)2C6H6
Quantity 1 g
Fieser 12,144; 16,99
Solubility Information Solubility in water: .. Other solubilities: soluble in benzene,toluene
Formula Weight 503.33
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protecti
EINECSNumber 255-686-6

RUO – Research Use Only