missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) thiophene-2-carboxylate

Catalog No. p-7056178
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6495003 1 g
AAH6495006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6495003 Supplier Thermo Scientific Chemicals Supplier No. H6495003
Only null left

CAS: 68986-76-5 | C5H3CuO2S | 190.683 g/mol

Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 68986-76-5
Molecular Formula C5H3CuO2S
Molecular Weight (g/mol) 190.683
MDL Number MFCD02183524
InChI Key SFJMFSWCBVEHBA-UHFFFAOYSA-M
Synonym copper i thiophene-2-carboxylate, cutc, unii-7519wbl07l, copper i thiophenecarboxylate, copper 2-thienylcarboxylate, cuprous 2-thienylcarboxylate, copper 2-thiophenecarboxylate, copper 1+ thiophene-2-carboxylate, copper i thiophene-2-carboxylate mi
PubChem CID 11194830
IUPAC Name copper(1+);thiophene-2-carboxylate
SMILES C1=CSC(=C1)C(=O)[O-].[Cu+]

Specifications

Color Brown to Red
Physical Form Powder
Quantity 1 g
Formula Weight 190.68
Sensitivity Air and light sensitive
Chemical Name or Material Copper(I) thiophene-2-carboxylate
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only